Learn More
MSC SPI Pin Gage Handle, 0.011-0.06 in, M0, Aluminum, 1 ea
Catalog No. 501920236
Encompass
This pin gage handle is designed for a ring diameter range of 0.011 to 0.06 in. It features an M0 handle size and is constructed from anodized aluminum, with an overall length of 5.20 in.
  • Functions like a drill chuck with three jaws
  • Holds any pin diameter within its advertised range
  • No bushings required for different pin sizes
  • Used to hold pin gages in go/no-go configurations
  • Provides easier handling and organization for pin gages
Catalog No. 50-192-0236 Supplier MSC Supplier No. 69814721
This pin gage handle is designed for a ring diameter range of 0.011 to 0.06 in. It features an M0 handle size and is constructed from anodized aluminum, with an overall length of 5.20 in.
  • Functions like a drill chuck with three jaws
  • Holds any pin diameter within its advertised range
  • No bushings required for different pin sizes
  • Used to hold pin gages in go/no-go configurations
  • Provides easier handling and organization for pin gages