Learn More
MSC SPI 13-312-4 Ring & Plug Thread Gage Holders & Handles; Type: Pin Gage Handle ; Handle Size: M0 ; Minimum Ring Diameter (Decimal Inch): 0.0610 ; Maximum Ring Diameter (Decimal Inch): 0.2500 ; Material: Aluminum
Supplier:  MSC 69814713
Encompass
SPI 13-312-4 Ring & Plug Thread Gage Holders & Handles; Type: Pin Gage Handle ; Handle Size: M0 ; Minimum Ring Diameter (Decimal Inch): 0.0610 ; Maximum Ring Diameter (Decimal Inch): 0.2500 ; Material: Aluminum
Catalog No. 50-192-0237