Learn More
MSC SPI Pin Gage Handle, 0.251-0.5 in, M2, Aluminum, 1 ea
Catalog No. 501920238
Encompass
This pin gage handle is constructed from aluminum and designed for use with pin gages, offering a secure grip through a three-jaw tightening mechanism. It accommodates pin gages within a specific diameter range, making it suitable for precise measurement tasks.
  • For pin gages
  • M2 handle size
  • Anodized coating
  • Features three jaws that tighten like a drill chuck
  • Overall length 6.20 in
  • Supports 0.251–0.5 in ring diameter range
Catalog No. 50-192-0238 Supplier MSC Supplier No. 69814705
This pin gage handle is constructed from aluminum and designed for use with pin gages, offering a secure grip through a three-jaw tightening mechanism. It accommodates pin gages within a specific diameter range, making it suitable for precise measurement tasks.
  • For pin gages
  • M2 handle size
  • Anodized coating
  • Features three jaws that tighten like a drill chuck
  • Overall length 6.20 in
  • Supports 0.251–0.5 in ring diameter range