Learn More
MSC SPI Handle, 0.501-0.625 in Range, M3 Size, Aluminum, 1 ea
Catalog No. 501920239
Encompass
This pin gage handle is designed to securely hold gage pins for various measurement applications. It accommodates pins within a specific diameter range, providing reliable performance for inspection tasks.
  • Accommodates pin diameters from 0.472 in (12.00 mm) to 0.650 in (16.50 mm)
  • M3 handle size for precise control
  • Constructed from durable aluminum with an anodized coating
  • Features an aluminum nut and metal spring-loaded plastic bushing
  • No additional bushings required for holding gage pins from 0.501 in to 0.625 in
  • Overall length of 6.00 in
Catalog No. 50-192-0239 Supplier MSC Supplier No. 69814689
This pin gage handle is designed to securely hold gage pins for various measurement applications. It accommodates pins within a specific diameter range, providing reliable performance for inspection tasks.
  • Accommodates pin diameters from 0.472 in (12.00 mm) to 0.650 in (16.50 mm)
  • M3 handle size for precise control
  • Constructed from durable aluminum with an anodized coating
  • Features an aluminum nut and metal spring-loaded plastic bushing
  • No additional bushings required for holding gage pins from 0.501 in to 0.625 in
  • Overall length of 6.00 in