Learn More
MSC SPI Pin Gage Handle, 0.917-1 in, M7, Aluminum, 1 ea
Catalog No. 501920242
Encompass
This pin gage handle is designed for precise measurement applications, accommodating pin gages with a ring diameter range of 0.917 to 1 " Constructed from aluminum, it provides a lightweight yet durable solution for laboratory and industrial use.
  • Chuck type holder, not requiring bushings
  • M7 handle size for pin gages
  • Accommodates ring diameters from 0.917 in to 1 in
  • Anodized aluminum construction for durability
  • Overall length of 6.20 in
Catalog No. 50-192-0242 Supplier MSC Supplier No. 69814606
This pin gage handle is designed for precise measurement applications, accommodating pin gages with a ring diameter range of 0.917 to 1 " Constructed from aluminum, it provides a lightweight yet durable solution for laboratory and industrial use.
  • Chuck type holder, not requiring bushings
  • M7 handle size for pin gages
  • Accommodates ring diameters from 0.917 in to 1 in
  • Anodized aluminum construction for durability
  • Overall length of 6.20 in