Learn More
MSC SPI Pin Gage Handle, M4, 0.626-0.75 Ring, Aluminum, 1 ea
Catalog No. 501920240
Encompass
The SPI pin gage handle is designed for use with pin gages. It features an M4 handle size and is made from anodized aluminum, suitable for precise measurement applications.
  • Made from anodized aluminum for durability
  • Features an M4 handle size for comfortable grip
  • Accommodates pin gages with a ring diameter range of 0.626–0.75 in
  • Overall length of 6.00 in
Catalog No. 50-192-0240 Supplier MSC Supplier No. 69814697
The SPI pin gage handle is designed for use with pin gages. It features an M4 handle size and is made from anodized aluminum, suitable for precise measurement applications.
  • Made from anodized aluminum for durability
  • Features an M4 handle size for comfortable grip
  • Accommodates pin gages with a ring diameter range of 0.626–0.75 in
  • Overall length of 6.00 in