Learn More
MSC SPI Pin Gage Handle, 0.751-0.832 in Ring Dia, 1 ea
Catalog No. 501920241
Encompass
This pin gage handle is designed for precise measurement applications, securely holding pin gages for accurate dimensional inspection. It is constructed from durable aluminum with an anodized finish for enhanced longevity.
  • Constructed from aluminum for durability
  • Anodized coating provides enhanced wear resistance
  • M5 handle size for compatibility with various pin gages
  • Accommodates pin gages with a ring diameter range of 0.751-0.832 in
  • Overall length of 6.20 in for comfortable handling
Catalog No. 50-192-0241 Supplier MSC Supplier No. 69814630
This pin gage handle is designed for precise measurement applications, securely holding pin gages for accurate dimensional inspection. It is constructed from durable aluminum with an anodized finish for enhanced longevity.
  • Constructed from aluminum for durability
  • Anodized coating provides enhanced wear resistance
  • M5 handle size for compatibility with various pin gages
  • Accommodates pin gages with a ring diameter range of 0.751-0.832 in
  • Overall length of 6.20 in for comfortable handling