Learn More
Mitegen Pin Lid Manipulator Tool By Nanosoft, Designed For Use With The Nanosoft Pin Lid For Hybrid Grid Box
SDP
Catalog No. NC2752341
Encompass
This Pin Lid Manipulator Tool is engineered for secure and precise handling of pin lids on cryo-EM grid boxes, enhancing safety and efficiency in your laboratory workflow. It is designed to work seamlessly with Hybrid Grid Box lids (SKU: M-CEM-NS-14151026) and other compatible grid box lids. The tool provides a robust, firm hold on pin lids, thanks to its specialized tip with integrated slots that offer controlled flex, ensuring a consistently tighter grip during manipulation. This innovative design offers several key advantages: it prevents fingers from freezing during cryogenic transfers, eliminates the issue of the tool unscrewing during use, and safeguards pin lids from cracking due to excessive pressure. By providing a dependable and ergonomic solution, this manipulator tool minimizes the risk of sample contamination or damage, contributing to reliable cryo-EM sample preparation and improved operational comfort for researchers.
Catalog No. NC2752341 Supplier Mitegen Supplier No. MCEMNSPLT01
This Pin Lid Manipulator Tool is engineered for secure and precise handling of pin lids on cryo-EM grid boxes, enhancing safety and efficiency in your laboratory workflow. It is designed to work seamlessly with Hybrid Grid Box lids (SKU: M-CEM-NS-14151026) and other compatible grid box lids. The tool provides a robust, firm hold on pin lids, thanks to its specialized tip with integrated slots that offer controlled flex, ensuring a consistently tighter grip during manipulation. This innovative design offers several key advantages: it prevents fingers from freezing during cryogenic transfers, eliminates the issue of the tool unscrewing during use, and safeguards pin lids from cracking due to excessive pressure. By providing a dependable and ergonomic solution, this manipulator tool minimizes the risk of sample contamination or damage, contributing to reliable cryo-EM sample preparation and improved operational comfort for researchers.