Learn More
Fritsch Milling And Sizing Inc Pin Mill Conversion Kit Cpl. Pin Insert, P-14 Classic Line
SDP
Catalog No. 501533942
Encompass
Pin mill conversion kit cpl. pin insert, PULVERISETTE 14 classic line
Consisting of pin rotor and grinding chamber lid with pin insert.
For grinding without a sieve for the most difficult, medium-hard, oil or grease containing materials such as waxes or paraffins.
To discharge and collect large quantities which need to be ground use the conversion kit for grinding large quantities.
Catalog No. 50-153-3942 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.2600.00
Pin mill conversion kit cpl. pin insert, PULVERISETTE 14 classic line
Consisting of pin rotor and grinding chamber lid with pin insert.
For grinding without a sieve for the most difficult, medium-hard, oil or grease containing materials such as waxes or paraffins.
To discharge and collect large quantities which need to be ground use the conversion kit for grinding large quantities.