Learn More
Waters Corp Waters Pin Plug, 1/16 in., High Pressure
Catalog No. 502039449 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pin plug is specifically designed for use in port 6 of the Trap Valve pod assembly within various nanoACQUITY system configurations. It is crucial for maintaining proper function in Direct Injection, Single Pump Trapping, Vented Dual Pump Trapping, and 2D with Dilution setups.
  • Required for port 6 of the Trap Valve pod assembly.
  • Supports nanoACQUITY configurations including Direct Injection, Single Pump Trapping, Vented Dual Pump Trapping, and 2D with Dilution.
  • Supplied in packages of 5 units.
  • Replaced as needed to ensure system integrity.
Catalog No. 50-203-9449 Supplier Waters Corp Supplier No. 700002747
This pin plug is specifically designed for use in port 6 of the Trap Valve pod assembly within various nanoACQUITY system configurations. It is crucial for maintaining proper function in Direct Injection, Single Pump Trapping, Vented Dual Pump Trapping, and 2D with Dilution setups.
  • Required for port 6 of the Trap Valve pod assembly.
  • Supports nanoACQUITY configurations including Direct Injection, Single Pump Trapping, Vented Dual Pump Trapping, and 2D with Dilution.
  • Supplied in packages of 5 units.
  • Replaced as needed to ensure system integrity.