Learn More
Waters Corp Pin Plug Assembly, Stainless Steel, 2/pk, HPLC System
Catalog No. 502390463 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement pin plug assembly is typically used in Autopurification Systems, including CFO or SFO modules. It is designed to block unused ports on Rheodyne Valves.
  • Made from stainless steel.
  • Supplied as 2 units per package.
  • Functions as a fitting.
  • Compatible with HPLC systems.
  • Replace as required.
Catalog No. 50-239-0463 Supplier Waters Corp Supplier No. 700001586
This replacement pin plug assembly is typically used in Autopurification Systems, including CFO or SFO modules. It is designed to block unused ports on Rheodyne Valves.
  • Made from stainless steel.
  • Supplied as 2 units per package.
  • Functions as a fitting.
  • Compatible with HPLC systems.
  • Replace as required.