Learn More
Zymo Research Corporation Pinpoint™ Slide DNA Isolation System (50 Preps) w/ Zymo-Spin™ I Columns (Uncapped)
SDP
Catalog No. NC9180999
Encompass_Preferred
The system combines our powerful Pinpoint tissuesampling technology with a one-step DNA extraction method and makes targeted tissue genomic DNA isolation easy and simple. The kit efficiently recovers genomic DNA from paraffin archives or fresh tissue sections for PCR analysis. Unlike the UV-fractionation method, the Pinpoint system does not destroy the surroundings of target cells. There is no need for expensive specialized equipment or computer software. Highlights- Easily isolates genomic DNA in any targeted microscopic tissue area on a slide.- The simple procedure combines Pinpoint tissue sampling technology with a one-step DNA extraction method.- Efficiently recovers genomic DNA from paraffin archives or fresh tissue sections for PCR analysis.
Catalog No. NC9180999 Supplier Zymo Research Corporation Supplier No. D3001
The system combines our powerful Pinpoint tissuesampling technology with a one-step DNA extraction method and makes targeted tissue genomic DNA isolation easy and simple. The kit efficiently recovers genomic DNA from paraffin archives or fresh tissue sections for PCR analysis. Unlike the UV-fractionation method, the Pinpoint system does not destroy the surroundings of target cells. There is no need for expensive specialized equipment or computer software. Highlights- Easily isolates genomic DNA in any targeted microscopic tissue area on a slide.- The simple procedure combines Pinpoint tissue sampling technology with a one-step DNA extraction method.- Efficiently recovers genomic DNA from paraffin archives or fresh tissue sections for PCR analysis.