Learn More
Description
This recombinant protein can be used for biological assays. For research use only.
Specifications
Specifications
| Accession Number | P63167 |
| For Use With (Application) | Not Available |
| Gene ID (Entrez) | 8655 |
| Molecular Weight (g/mol) | 12.5 kDa |
| Quantity | 0.05 mg |
| Source | E. coli |
| Common Name | dynein light chain LC8-type 1 |
| Gene Symbol | DYNLL1 |
| Product Type | Recombinant Protein |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
