Learn More
Electron Microscopy Sciences SEM Pin Stub Adapter, 38 mm x 15 mm
SDP
Catalog No. 5027868
Encompass
Used to adapt 1/8" (3.1mm) pin diameter SEM stubs. Made from vacuum grade aluminum with a stainless steel set screw. Allen wrench included.
Diameter: 38 mm
Height: 15 mm
Catalog No. 50-278-68 Supplier Electron Microscopy Sciences Supplier No. 7594015
Used to adapt 1/8" (3.1mm) pin diameter SEM stubs. Made from vacuum grade aluminum with a stainless steel set screw. Allen wrench included.
Diameter: 38 mm
Height: 15 mm