Learn More
Electron Microscopy Sciences SEM Pin Stub Adapter, 10 mm x 10 mm
SDP
Catalog No. 5027867
Encompass
Used to adapt 1/8" (3.1mm) pin diameter SEM stubs. Made from vacuum grade aluminum with a stainless steel set screw. Allen wrench included.
Diameter: 10 mm
Height: 10 mm
Catalog No. 50-278-67 Supplier Electron Microscopy Sciences Supplier No. 7594010
Used to adapt 1/8" (3.1mm) pin diameter SEM stubs. Made from vacuum grade aluminum with a stainless steel set screw. Allen wrench included.
Diameter: 10 mm
Height: 10 mm