Learn More
Ted Pella Inc SEM Pin Stub Mount Gripper Tweezers, SS, 45° angle, 114mm long, for 12.7mm pin stubs
SDP
Catalog No. NC9828398
Encompass
SEM Mount Tweezers for handling grooved pin stubs. Conveniently grasp the grooved head of SEM pin stubs with 12.7mm diameter. Stainless Steel, 45° angle, 114mm long.
Catalog No. NC9828398 Supplier Ted Pella Inc Supplier No. 166312
SEM Mount Tweezers for handling grooved pin stubs. Conveniently grasp the grooved head of SEM pin stubs with 12.7mm diameter. Stainless Steel, 45° angle, 114mm long.