Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 2" Head, Aluminum, 3.2 mm Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 7563650
Encompass_Preferred
For AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, or Tescan. Made from lathe finished ultra-pure aluminum.
Head: 2"
Pin diameter: 3.2 mm
Catalog No. 50-190-1450