Learn More
Electron Microscopy Sciences Gatan 3View SEM Pin Stubs Standard, 1.4 mm Head, 2 x 12.5 mm Pin
SDP
Supplier:  Electron Microscopy Sciences 7595902
Encompass
The Gatan 3View systems use special sample pins. These are used inside SEMs to create 3D imaging data of samples by serial slicing and BSD imaging. These pins are fully compatible with the Gatan 3View systems; 2 mm pin, 3 mm head with cone top and total height of 12.5 mm. Manufactured from vacuum grade aluminium.
Head: 1.4 mm
Pin: 2 x 12.5 mm
Catalog No. 50-192-9103