Learn More
Cayman Chemical PIn Thromboxn A2 500ug
SDP
Catalog No. 502982077
Encompass_Preferred
PTA2 is a stable analog of TXA2. It is a TP receptor antagonist and an inhibitor of TX synthase.{516,794} PTA2 inhibits U-46619-induced cat coronary artery constriction (ID50 = 0.1 µM), U-46619-induced aggregation of human platelets (IC50 = 2 µM), and rabbit platelet TX synthase (ID50 = 50 µM). PTA2 does not affect PGI synthase up to a concentration of 100 µM.{516}
Catalog No. 50-298-2077 Supplier Cayman Chemical Supplier No. 19020500 UG
PTA2 is a stable analog of TXA2. It is a TP receptor antagonist and an inhibitor of TX synthase.{516,794} PTA2 inhibits U-46619-induced cat coronary artery constriction (ID50 = 0.1 µM), U-46619-induced aggregation of human platelets (IC50 = 2 µM), and rabbit platelet TX synthase (ID50 = 50 µM). PTA2 does not affect PGI synthase up to a concentration of 100 µM.{516}