Learn More
Sino Biological Pin1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10282T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Pin1 (rh Pin1; Catalog#10282-H07E; Q13526-1; Met1-Glu163). Pin1 specific IgG was purified by Human Pin1 affinity chromatography.
Catalog No. 50-224-6304