Learn More
Medchemexpress LLC PIN1 inhibitor API-1 100mg | 680622-70-2 | 366.30 | C15H13F3N6O2 | 100 MG
SDP
Catalog No. 5000227364
Encompass_Preferred
PIN1 inhibitor API-1 is a small-molecule research compound that inhibits PIN1 activity and functions as a microRNA activator. It is supplied as a research-grade powder (100 mg) intended for preclinical in vitro and in vivo studies; consult the certificate of analysis and safety data sheet for storage, purity, and handling instructions.
  • High purity: documented at about 99.6%.
  • Molecular weight: 366.30; chemical formula: C15H13F3N6O2.
  • High DMSO solubility: up to 250 mg/mL with ultrasonic assistance.
  • Manufacturer provides in vivo formulation examples and solubility protocols.
  • Data sheet and safety data sheet available for characterization and safe handling.
Catalog No. 50-002-27364 Supplier Medchemexpress LLC Supplier No. HY116716100MG
PIN1 inhibitor API-1 is a small-molecule research compound that inhibits PIN1 activity and functions as a microRNA activator. It is supplied as a research-grade powder (100 mg) intended for preclinical in vitro and in vivo studies; consult the certificate of analysis and safety data sheet for storage, purity, and handling instructions.
  • High purity: documented at about 99.6%.
  • Molecular weight: 366.30; chemical formula: C15H13F3N6O2.
  • High DMSO solubility: up to 250 mg/mL with ultrasonic assistance.
  • Manufacturer provides in vivo formulation examples and solubility protocols.
  • Data sheet and safety data sheet available for characterization and safe handling.