Learn More
Medchemexpress LLC PIN1 inhibitor API-1 25mg | 680622-70-2 | 366.30 g/mol | C15H13F3N6O2 | 25 MG
SDP
Catalog No. 5000228110
Encompass_Preferred
PIN1 inhibitor API-1 is a small-molecule research compound that inhibits Pin1 (peptidyl-prolyl cis-trans isomerase NIMA-interacting 1) and functions as a microRNA activator. Supplied as a solid with high reported purity, it is suitable for in vitro biochemical and cell-based studies and is soluble in DMSO and in certain co-solvent formulations.
Catalog No. 50-002-28110 Supplier Medchemexpress LLC Supplier No. HY11671625MG
PIN1 inhibitor API-1 is a small-molecule research compound that inhibits Pin1 (peptidyl-prolyl cis-trans isomerase NIMA-interacting 1) and functions as a microRNA activator. Supplied as a solid with high reported purity, it is suitable for in vitro biochemical and cell-based studies and is soluble in DMSO and in certain co-solvent formulations.