Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PIN1 inhibitor API-1 50mg | 680622-70-2 | 366.30 | C15H13F3N6O2 | 50 MG
SDP

Catalog No. 5000227388
Encompass_Preferred

PIN1 inhibitor API-1 is a selective peptidyl-prolyl cis-trans isomerase Pin1 inhibitor (IC50 = 72.3 nM) that binds the Pin1 PPIase domain and has been reported to up-regulate anticancer microRNA biogenesis and suppress hepatocellular carcinoma in preclinical models.

  • Selective Pin1 inhibitor (IC50 72.3 nM).
  • Molecular formula C15H13F3N6O2; molecular weight 366.30.
  • Purity 99.63%.
  • Appearance solid; color white to yellow.
  • Soluble in DMSO (250 mg/mL); in vivo formulations ≥ 2.17 mg/mL in mixed solvents.
  • Available in multiple package sizes including 50 MG.

Catalog No. 50-002-27388 Supplier Medchemexpress LLC Supplier No. HY11671650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PIN1 inhibitor API-1 is a selective peptidyl-prolyl cis-trans isomerase Pin1 inhibitor (IC50 = 72.3 nM) that binds the Pin1 PPIase domain and has been reported to up-regulate anticancer microRNA biogenesis and suppress hepatocellular carcinoma in preclinical models.

  • Selective Pin1 inhibitor (IC50 72.3 nM).
  • Molecular formula C15H13F3N6O2; molecular weight 366.30.
  • Purity 99.63%.
  • Appearance solid; color white to yellow.
  • Soluble in DMSO (250 mg/mL); in vivo formulations ≥ 2.17 mg/mL in mixed solvents.
  • Available in multiple package sizes including 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.