Learn More
Selleck Chemical LLC PIN1 inhibitor API-1
SDP
Supplier:  Selleck Chemical LLC S6893100MG
Encompass_Preferred
PIN1 inhibitor API-1 is a specific Pin1 (peptidyl-prolyl cis-trans isomerase NIMA-interacting 1) inhibitor with IC50 of 72 3 nM PIN1 inhibitor API-1 targets Pin1 peptidyl-prolyl isomerase domain and inhibits Pin1 cis-trans isomerizing activity PIN1 inhibitor API-1 up-regulates miRNA biogenesis by retaining active XPO5 conformation and suppresses HCC development
Catalog No. 50-000-02635