Learn More
Enzo Life Sciences Pinacidil . monohydrate (50mg). CAS: 85371-64-8
SDP
Supplier:  Enzo Life Sciences ALX550285M050
Encompass_Preferred
K+ channel activator. Antihypertensive. Alternative name: (+/-)-N-Cyano-N-4-pyridinyl-N-(1,2,2-trimethylpropyl)guanidine monohydrate. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (14mg/ml), DMSO (28mg/ml) or dilute aqueous acid; insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9525