Learn More
Cayman Chemical Pinacidil (hydrate), Purity: ≥95%, CAS: 85371-64-8, 100 mg
SDP
Catalog No. 502971100
Encompass_Preferred
A potassium channel opener, activating the ATP-modulated potassium channels of guinea pig bladder and heart with Ki values of 104 and 251 nM, respectively; completely relaxes coronary artery rings preconstricted with serotonin (IC50 = 1.26 µM)
Catalog No. 50-297-1100 Supplier Cayman Chemical Supplier No. 15416100 MG
A potassium channel opener, activating the ATP-modulated potassium channels of guinea pig bladder and heart with Ki values of 104 and 251 nM, respectively; completely relaxes coronary artery rings preconstricted with serotonin (IC50 = 1.26 µM)