Learn More
Cayman Chemical Pinacidil (hydrate), Purity: ≥95%, CAS: 85371-64-8, 250 mg
SDP
Catalog No. 502969121
Encompass_Preferred
A potassium channel opener, activating the ATP-modulated potassium channels of guinea pig bladder and heart with Ki values of 104 and 251 nM, respectively; completely relaxes coronary artery rings preconstricted with serotonin (IC50 = 1.26 µM)
Catalog No. 50-296-9121 Supplier Cayman Chemical Supplier No. 15416250 MG
A potassium channel opener, activating the ATP-modulated potassium channels of guinea pig bladder and heart with Ki values of 104 and 251 nM, respectively; completely relaxes coronary artery rings preconstricted with serotonin (IC50 = 1.26 µM)