Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pinacol (Dichloromethyl) Boronate

Catalog No. 9010625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-106-25G 1 g
90-106-21G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-106-25G Supplier Sigma Aldrich Supplier No. 9010625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pinacol (dichloromethyl) boronate is a general reagent that was employed by Prof. Greg C. Fu and coworkers to achieve the iterative synthesis of asymmetric (secondary) alkyl boronate esters. These can be further derivatized through a growing suite of chemical transformations developed in the last decade (cross coupling, oxidation, amination, halogenation) that proceed with stereochemical fidelity.A general, modular method for the catalytic asymmetric synthesis of alkylboronate esters

Specifications

CAS 83622-41-7
Density 1.1423 g/mL
Molecular Formula C7H13BCl2O2
Refractive Index n/D 1.4511
Linear Formula C7H13BCl2O2
MDL Number MFCD27976342
Quantity 1 g
Synonym 1,3,2-Dioxaborolane, 2-(dichloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; Dichloromethyl pinacol boronate
Molecular Weight (g/mol) 210.89 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.