Learn More
Medchemexpress LLC (+)-Pinanediol | 18680-27-8 | MFCD00077851 | 99.98% | 170.25 | 25 G
SDP
Catalog No. 5000402841
Encompass_Preferred
(+)-Pinanediol is a potent inducer of melanin production. In S91 cells, it induces higher levels of pigmentation and nitric oxide (NO) synthesis. This compound is an effective inducer of melanogenesis.
  • Potent inducer of melanin production
  • Induces higher levels of pigmentation in S91 cells
  • Induces nitric oxide (NO) synthesis in S91 cells
  • Effective inducer of melanogenesis
  • For research use only
Catalog No. 50-004-02841 Supplier Medchemexpress LLC Supplier No. 25GHY20074
Add to Cart
Add to Cart
(+)-Pinanediol is a potent inducer of melanin production. In S91 cells, it induces higher levels of pigmentation and nitric oxide (NO) synthesis. This compound is an effective inducer of melanogenesis.
  • Potent inducer of melanin production
  • Induces higher levels of pigmentation in S91 cells
  • Induces nitric oxide (NO) synthesis in S91 cells
  • Effective inducer of melanogenesis
  • For research use only