Learn More
Selleck Chemical LLC Pinaverium bromide
SDP
Catalog No. 5000004107
Encompass_Preferred
Pinaverium bromide (Icosapentaenoic acid) acts as a calcium channel blocker and is useful for functional gastrointestinal disorders
Catalog No. 50-000-04107 Supplier Selleck Chemical LLC Supplier No. S647325MG
Pinaverium bromide (Icosapentaenoic acid) acts as a calcium channel blocker and is useful for functional gastrointestinal disorders