Learn More
Selleck Chemical LLC Pinaverium bromide
SDP
Supplier:  Selleck Chemical LLC S6473100MG
Encompass_Preferred
Pinaverium bromide (Icosapentaenoic acid) acts as a calcium channel blocker and is useful for functional gastrointestinal disorders
Catalog No. 50-000-04644