Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinaverium bromide | 53251-94-8 | 99.7% | 591.42 | 25 MG
SDP

Catalog No. 5000439554
Encompass_Preferred

Pinaverium bromide | 53251-94-8 | 99.7% | 591.42 | 25 MG

Catalog No. 50-004-39554 Supplier Medchemexpress LLC Supplier No. 25MGHY111613
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinaverium bromide | 53251-94-8 | 99.7% | 591.42 | 25 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.