Learn More
Medchemexpress LLC Pinaverium bromide | 53251-94-8 | MFCD08063574 | 98.0% | 591.42 g/mol | C26H41Br2NO4 | 5 MG
SDP
Catalog No. 5000263401
Encompass_Preferred
Pinaverium bromide is an analytical research standard of an L-type calcium channel blocker with selectivity for the gastrointestinal tract. Supplied in small laboratory pack sizes, it is intended for analytical and pharmacological research, method development, and use as a reference material. The compound has formula C26H41Br2NO4, molecular weight 591.42 g/mol, and CAS 53251-94-8.
  • Research-grade analytical standard for L-type calcium channel studies.
  • Supplied in small, lab-friendly pack sizes for routine use.
  • High purity suitable for analytical and pharmacological assays.
  • Datasheet, safety data sheet, and certificate of analysis available on request.
  • Stable bromide salt form appropriate for gastrointestinal pharmacology research.
Catalog No. 50-002-63401 Supplier Medchemexpress LLC Supplier No. HY1116135MG
Pinaverium bromide is an analytical research standard of an L-type calcium channel blocker with selectivity for the gastrointestinal tract. Supplied in small laboratory pack sizes, it is intended for analytical and pharmacological research, method development, and use as a reference material. The compound has formula C26H41Br2NO4, molecular weight 591.42 g/mol, and CAS 53251-94-8.
  • Research-grade analytical standard for L-type calcium channel studies.
  • Supplied in small, lab-friendly pack sizes for routine use.
  • High purity suitable for analytical and pharmacological assays.
  • Datasheet, safety data sheet, and certificate of analysis available on request.
  • Stable bromide salt form appropriate for gastrointestinal pharmacology research.