Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinaverium impurity 1 | 54370-00-2 | MFCD02093896 | 247.09 | C9H11BrO3 | 100 MG
SDP

Catalog No. 5000283458
Encompass_Preferred

Pinaverium impurity 1 is a chemical impurity related to pinaverium, supplied for research and analytical use. It is provided as a drug intermediate and characterized by its molecular composition and identifiers.

  • Used as an impurity reference or intermediate in analytical and synthetic work.
  • Chemical formula C9H11BrO3, molecular weight 247.09.
  • CAS number 54370-00-2 for unambiguous identification.
  • Intended for research use only; not for human or animal therapeutic use.
  • Storage and handling instructions provided on the certificate of analysis.

Catalog No. 50-002-83458 Supplier Medchemexpress LLC Supplier No. HYW137686100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinaverium impurity 1 is a chemical impurity related to pinaverium, supplied for research and analytical use. It is provided as a drug intermediate and characterized by its molecular composition and identifiers.

  • Used as an impurity reference or intermediate in analytical and synthetic work.
  • Chemical formula C9H11BrO3, molecular weight 247.09.
  • CAS number 54370-00-2 for unambiguous identification.
  • Intended for research use only; not for human or animal therapeutic use.
  • Storage and handling instructions provided on the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.