Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinaverium impurity 1 | 54370-00-2 | MFCD02093896 | 247.09 | C9H11BrO3 | 5 G
SDP

Catalog No. 5000282713
Encompass_Preferred

Pinaverium impurity 1 is a characterized impurity of the L-type calcium channel blocker pinaverium, supplied for research and analytical use as a drug intermediate and reference material. It is intended for laboratory method development and impurity profiling and is not for human or clinical use.

  • Chemical identification: CAS 54370-00-2.
  • Chemical formula: C9H11BrO3.
  • Molecular weight: 247.09.
  • Intended use: research and analytical applications.
  • Pack quantity: 5 G.
  • Storage: follow conditions in the certificate of analysis.

Catalog No. 50-002-82713 Supplier Medchemexpress LLC Supplier No. HYW1376865G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinaverium impurity 1 is a characterized impurity of the L-type calcium channel blocker pinaverium, supplied for research and analytical use as a drug intermediate and reference material. It is intended for laboratory method development and impurity profiling and is not for human or clinical use.

  • Chemical identification: CAS 54370-00-2.
  • Chemical formula: C9H11BrO3.
  • Molecular weight: 247.09.
  • Intended use: research and analytical applications.
  • Pack quantity: 5 G.
  • Storage: follow conditions in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.