Learn More
Medchemexpress LLC Pinaverium impurity 1 | 54370-00-2 | MFCD02093896 | 96.5% | 247.09 g/mol | C9H11BrO3 | 250 MG
SDP
Catalog No. 5000282859
Encompass_Preferred
Pinaverium impurity 1 is a characterized impurity of pinaverium supplied as an analytical reference for laboratory research and method development. It is intended for research use only and is not for human or clinical use. The substance has chemical formula C9H11BrO3, CAS 54370-00-2, and a molecular weight of 247.09 g/mol. Store under conditions specified on the certificate of analysis.
  • Suitable as an analytical reference standard for impurity profiling and method validation.
  • Provides confirmed chemical identifiers (CAS, formula, molecular weight).
  • Intended for research use only, not for clinical applications.
  • Certificate of analysis available for storage and purity details.
  • Supplied for laboratory and analytical workflows.
Catalog No. 50-002-82859 Supplier Medchemexpress LLC Supplier No. HYW137686250MG
Pinaverium impurity 1 is a characterized impurity of pinaverium supplied as an analytical reference for laboratory research and method development. It is intended for research use only and is not for human or clinical use. The substance has chemical formula C9H11BrO3, CAS 54370-00-2, and a molecular weight of 247.09 g/mol. Store under conditions specified on the certificate of analysis.
  • Suitable as an analytical reference standard for impurity profiling and method validation.
  • Provides confirmed chemical identifiers (CAS, formula, molecular weight).
  • Intended for research use only, not for clinical applications.
  • Certificate of analysis available for storage and purity details.
  • Supplied for laboratory and analytical workflows.