Learn More
TARGETMOL CHEMICALS INC PINCAINIDE 5MG
SDP
Catalog No. 502576341
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Pincainide is a novel beta-aminoaniline that inhibits in a dose-dependent manner diseases caused by abnormal norepinephrine vascular smooth muscle function. purity: 99%
Catalog No. 50-257-6341 Supplier TARGETMOL CHEMICALS INC Supplier No. T717465MG
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Pincainide is a novel beta-aminoaniline that inhibits in a dose-dependent manner diseases caused by abnormal norepinephrine vascular smooth muscle function. purity: 99%