Learn More
Qosina Corp PINCH CLAMP NON-REOPEN .25-
Catalog No. NC2380186
Encompass
Pinch Clamp, Non-reopening, NaturalMaterials: PPFits 0.25 inch - 0.315 inch OD Tubing (6.4 mm - 8 mm)
Catalog No. NC2380186 Supplier Qosina Corp Supplier No. 13991
Pinch Clamp, Non-reopening, NaturalMaterials: PPFits 0.25 inch - 0.315 inch OD Tubing (6.4 mm - 8 mm)