Learn More
Chemglass Life Sciences Clamp, Pinch, Stainless Steel, #75
SDP
Catalog No. CG15021
Encompass_Preferred
Clamp, Pinch, Stainless Steel, #75. Made entirely of stainless steel. Part numbers CG-150-01 and CG-150-03 are supplied WITHOUT a screw type locking device. All other sizes have the screw type locking device.Note: The size #102 Stainless Steel pinch clamp is available on a special order basis.
Catalog No. CG15021 Supplier Chemglass Life Sciences Supplier No. CG15021
Clamp, Pinch, Stainless Steel, #75. Made entirely of stainless steel. Part numbers CG-150-01 and CG-150-03 are supplied WITHOUT a screw type locking device. All other sizes have the screw type locking device.Note: The size #102 Stainless Steel pinch clamp is available on a special order basis.