Learn More
BASECAMP PROCESS COMPONENTS LLC PINCH VALV TYP VMC, 316 1"SS
Catalog No. NC2282489
Encompass
Pinch Valve, Type VMC, 1", direct air operated,tri-clamp,Body: 316 stainless steel;
This item is not returnable. View return policy
Catalog No. NC2282489 Supplier BASECAMP PROCESS COMPONENTS LLC Supplier No. VMC25.02XK.50T.50
Pinch Valve, Type VMC, 1", direct air operated,tri-clamp,Body: 316 stainless steel;