Learn More
Waters Corp Pinch Valve Assembly, with Fittings, MS System, Replacement
Catalog No. 502392410 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Pinch Valve Assembly is a replacement component designed with fittings for mass spectrometry systems. It is specifically compatible with QDa instruments featuring a KAB serial prefix and is positioned externally to the instrument.
  • Situated outside the system.
  • Designed as a replacement part.
  • Includes necessary fittings.
  • Compatible with MS systems.
  • For QDa instruments with KAB serial prefix.
  • Replacement recommended as required.
Catalog No. 50-239-2410 Supplier Waters Corp Supplier No. 700010360
This Pinch Valve Assembly is a replacement component designed with fittings for mass spectrometry systems. It is specifically compatible with QDa instruments featuring a KAB serial prefix and is positioned externally to the instrument.
  • Situated outside the system.
  • Designed as a replacement part.
  • Includes necessary fittings.
  • Compatible with MS systems.
  • For QDa instruments with KAB serial prefix.
  • Replacement recommended as required.