Learn More
Essendant PINE SOL 80 OZ
Supplier:  Essendant CLO60606CT
Encompass_Preferred
With Pine-Sol Multi-Surface Cleaner Disinfectant Concentrated from CloroxPro™, you only need one product to fight tough dirt, grease and grime. A 2X concentrated formula delivers more cleaning power per drop than previous Pine-Sol Pine formula. Easier to handle and easier to store, the space-saving bottle gives you more cleaning solution in a smaller bottle made of 30% less plastic. This multipurpose cleaner cleans, deodorizes and disinfects in one step, killing 99.9% of germs and leaving a classic scent. It cuts through grime and soap scum and is safe for use on wood, tile, stone, glass and more when used as directed on hard nonporous surfaces. Do not use on marble, aluminum, copper or wood that is unfinished, unsealed, unpainted, waxed, oiled or worn. Application: Cleaner/Disinfectant; Applicable Material: Baked Enamel; Bamboo; Cement; Chrome; Concrete; [Engineered] Wood; Glazed Ceramic Tiles; Glazed Porcelain; [Hard] Plastic; Hard Vinyl; Laminate; Linoleum; Metal,
Catalog No. NC2881274