Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Pinene, 97%, stabilized with alpha-Tocopherol

Catalog No. p-184566
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164360050 5 g Glass bottle
AC164362500 250 g Glass bottle
AC164360010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC164360050 Supplier Thermo Scientific Chemicals Supplier No. 164360050
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 80-56-8 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-56-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001339
InChI Key GRWFGVWFFZKLTI-UHFFFAOYSA-N
Synonym alpha-pinene, 2-pinene, .alpha.-pinene, acintene a, pinene isomer, 2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, pinene, alpha, pin-2 3-ene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl, +-a-pinene
PubChem CID 6654
ChEBI CHEBI:36740
IUPAC Name 4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CCC2CC1C2(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -55.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 154.0°C to 157.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 5 g
Specific Gravity 0.85
Merck Index 15, 7556
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
Stabilizer 0.8% alpha-tocopherol
Viscosity <7 mm2/s (40°C)
Formula Weight 136.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material α-Pinene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful if swallowed.
Very toxic to aquatic life with long lasting
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off imm
EINECSNumber 201-291-9
RTECSNumber DT7000000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.