Learn More
StatLab Pinfed Slide Labels
SDP
Catalog No. 21010343
Change view
Click to view available options
Type:
Slide Label
Catalog No. Type
21-010-343 Slide Label
1 options
Catalog No. 21-010-343 Supplier StatLab Supplier No. MLPF6/CS
Add to Cart
Add to Cart
Fan-folded and perforated between each row.
  • Die-cut 15/16 in. x 15/16 in., 6-across
  • Carrier width 7 in.
  • Packaged 25,056/Cs.
  • Fan-folded and perforated between each row
Specifications
Type Slide Label