Learn More
Electron Microscopy Sciences Pinhole Aperture, 1000 µm
SDP
Catalog No. 5033129
Encompass_Preferred
These pinholes are prepared from pure copper foil, 3 mm in diameter, 25 microns thick. They possess very high roundness and edge retention. Blackened on one surface. Mounted in black anodized aluminum discs. Mounted in a recessed hole in an anodized holder, 16 mm in diameter.
Catalog No. 50-331-29 Supplier Electron Microscopy Sciences Supplier No. PHC1000
These pinholes are prepared from pure copper foil, 3 mm in diameter, 25 microns thick. They possess very high roundness and edge retention. Blackened on one surface. Mounted in black anodized aluminum discs. Mounted in a recessed hole in an anodized holder, 16 mm in diameter.