Learn More
Footwear at Work PINK 6 INCH SAFETY TOE WMNS
Catalog No. NC1895534
Encompass_Preferred
The incredibly durable and versatile Alice women’s work book by Moxie Trades features a steel toe safety toe and a puncture-resistant plate for use in heavy-duty environments. The boot is constructed with a waterproof membrane and a full-grain, nubuck upper and a direct-attach PU outsole, giving you the comfort you need for long days.ASTM F2413-18 steel safety toeFull-grain, soft nubuck leatherPuncture-resisting plateSlip and oil-resisting outsoleMemory foam insoleExtra black lace in boxASTM F2413-18 electrical hazard, PRCSA Approved Grade 1.
Catalog No. NC1895534 Supplier Footwear at Work Supplier No. MT50162
The incredibly durable and versatile Alice women’s work book by Moxie Trades features a steel toe safety toe and a puncture-resistant plate for use in heavy-duty environments. The boot is constructed with a waterproof membrane and a full-grain, nubuck upper and a direct-attach PU outsole, giving you the comfort you need for long days.ASTM F2413-18 steel safety toeFull-grain, soft nubuck leatherPuncture-resisting plateSlip and oil-resisting outsoleMemory foam insoleExtra black lace in boxASTM F2413-18 electrical hazard, PRCSA Approved Grade 1.