Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ Pink PIPETMAN™ L

Catalog No. FA10006PNKG Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
100 to 1000 μL
2 to 20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
FA10006PNKG 100 to 1000 μL
FA10003PNKG 2 to 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FA10006PNKG Supplier Gilson™ Supplier No. FA10006PINK

With each purchase of the limited edition Pink PIPETMAN™ L, Gilson™ will donate a portion of the proceeds to a cancer research institute. It is designed with a natural fit that allows you to relax your grip, which reduces fatigue when pipetting.

  • Optimized comfort, safety, precision, and traceability
  • Offer an ideal weight and handle shape
  • Air-displacement-based limited edition
  • To decrease the risk of repetitive strain injuries, the PIPETMAN™ L features an updated piston assembly with more responsive springs, which reduces pipetting forces
  • The tip ejector is also adjustable for right- or left-handedness, whichever is most comfortable for you
  • Offers a true volume-locking mechanism to guarantee no accidental volume change during pipetting cycles, which could alter accuracy and precision of your results
  • The patented locking system fully disconnects the operating rod from the counter assembly during pipetting so that once locked into place, the volume cannot be accidentally changed, preventing accidental volume drift during pipetting tasks
  • To help you minimize cross-contamination, PIPETMAN™ L is fully autoclavable with no disassembly required for maximum convenience
  • Has a 2D identification bar code for easy traceability of the product part number, date of manufacturing, and serial number
  • Using a universal scanner, the pipette information can be downloaded to a spreadsheet or other software for tracking, which is particularly suited for enhanced GLP compliance
  • Has a color-coded push button that corresponds in color to Gilson™ PIPETMAN™ Tip packages
  • With each purchase of the limited edition pink PIPETMAN™ L, Gilson™ will donate a portion of the proceeds to a cancer research institute

Specifications

Compatible Tips Universal
Autoclavable Autoclavable
Color Pink
For Use With Pipette Tip
Material Polypropylene
Model P1000L
Number of Channels 1
Product Line Pipetman™ L
Type Variable Volume Air Displacement Pipette
Volume (Metric) 100 to 1000 μL
For Use With (Equipment) Pipette Tips
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.