Learn More
USA Scientific Inc Tough-Spots, 3/8 inch diameter, pink, on rolls
Catalog No. NC9236768
Encompass
Tough-Spots, small, pink labels for 0.5 mL tubes, 3/8 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. NC9236768 Supplier USA Scientific Inc Supplier No. 91850007
Tough-Spots, small, pink labels for 0.5 mL tubes, 3/8 inch diameter, 1000/roll. For -196C to +80C.