Learn More
Aviva Systems Biology Corp PINK1 Antibody - N-terminal region (ARP63313_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63313_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PINK1. It was validated on Western Blot. . Alternate names: BRPK, FLJ27236, PARK6ApplicationWB, IHC. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-936