Learn More
Biomatik Corporation PINK1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50429
Encompass_Preferred
PINK1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Pink1. The PINK1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, WB, IHC. Target Synonyms: Pink1. Accession Number: Q9BXM7. Gene ID: 65018. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6588